5-(Methoxycarbonyl)thiophene-3-boronic acid 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-boronothiophene-2-carboxylate, 4-Borono-2-(methoxycarbonyl)thiophene, (5-(Methoxycarbonyl)thiophen-3-yl)boronic acid

OR11695-1G
APOSOR11695-1GEA 265 EUR
APOSOR11695-1G
5-(Methoxycarbonyl)thiophene-3-boronic acid 96%
5-(Methoxycarbonyl)thiophene-3-boronic acid
Formel: C₆H₇BO₄S
Molecular Weight: 185.993 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09800878
CAS-Nummer: 957062-52-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR