2-Methoxy-5-(methoxycarbonyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

OR11696-1G OR11696-100MG
APOSOR11696-1GEA 197 EUR
APOSOR11696-1G APOSOR11696-100MG
2-Methoxy-5-(methoxycarbonyl)phenylboronsäure
2-Methoxy-5-(methoxycarbonyl)phenylboronsäure
Formel: C₉H₁₁BO₅
Lagertemperatur: Kühlschrank
MDL: MFCD09800877
CAS-Nummer: 221006-63-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR