5-(Ethoxycarbonyl)thiophene-3-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-(Ethoxycarbonyl)thiophen-3-yl)boronic acid, Ethyl 4-boronothiophene-2-carboxylate, 4-Borono-2-(ethoxycarbonyl)thiophene

OR11699-1G
APOSOR11699-1GEA 243 EUR
APOSOR11699-1G
5-(Ethoxycarbonyl)thiophene-3-boronic acid
5-(Ethoxycarbonyl)thiophene-3-boronic acid
Formel: C₇H₉BO₄S
Molecular Weight: 200.02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09800891
CAS-Nummer: 957121-19-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR