2-(Fur-2-yl)pyrrolidine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Pyrrolidin-2-yl)furan
OR11707-1G
APOSOR11707-1GEA
166
EUR
APOSOR11707-1G
2-(Fur-2-yl)pyrrolidine
2-(Fur-2-yl)pyrrolidine
Formel:
C₈H₁₁NO Molecular Weight: 137.179 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02663415 CAS-Nummer: 90086-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...