2-(Chloromethyl)-1-methylpiperidine hydrochloride

Lieferant: Apollo Scientific

OR11727-1G
APOSOR11727-1GEA 93 EUR
APOSOR11727-1G
2-(Chloromethyl)-1-methylpiperidine hydrochloride
2-(Chloromethyl)-1-methylpiperidine hydrochloride
Formel: C₇H₁₅Cl₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD09800596
CAS-Nummer: 27483-92-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR