3-Aminomethyl-3-hydroxymethyloxetane 90+%

Lieferant: Apollo Scientific
Gefahr

OR11732-1G
APOSOR11732-1GEA 0 EUR
APOSOR11732-1G
3-Aminomethyl-3-hydroxymethyloxetane 90+%
3-Aminomethyl-3-hydroxymethyloxetane
Formel: C5H11NO2
Lagertemperatur: Raumtemperatur
CAS-Nummer: 45513-32-4
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR