1H-Imidazol-2-amine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR11741-250MG
APOSOR11741-250MGEA 0 EUR
APOSOR11741-250MG
1H-Imidazol-2-amine hydrochloride
1H-Imidazol-2-amine hydrochloride
Formel: C₃H₆ClN₃
Molecular Weight: 119.55 g/mol
Schmelzpunkt: 152…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06795693
CAS-Nummer: 57575-96-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR