5-Chloro-2-hydroxy-4-methylbenzaldehyde 95%

Lieferant: Apollo Scientific

OR1185-1G
APOSOR1185-1GEA 81.2 EUR
APOSOR1185-1G
5-Chloro-2-hydroxy-4-methylbenzaldehyde 95%
5-Chloro-2-hydroxy-4-methylbenzaldehyde
Formel: C₈H₇ClO₂
Molecular Weight: 170,593 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094618
CAS-Nummer: 3328-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR