5-Brom-N-propylpicolinamid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-N-propylpyridine-2-carboxamide

OR11907-250MG
APOSOR11907-250MGEA 29 EUR
APOSOR11907-250MG
5-Brom-N-propylpicolinamid
5-Brom-N-propylpicolinamid
Formel: C₉H₁₁BrN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09801061
CAS-Nummer: 845305-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR