3-Bromo-2,4-dichlorobenzoic acid

Lieferant: Apollo Scientific
Achtung

OR11909-100MG
APOSOR11909-100MGEA 21.9 EUR
APOSOR11909-100MG
3-Bromo-2,4-dichlorobenzoic acid
3-Bromo-2,4-dichlorobenzoic acid
Formel: C₇H₃BrCl₂O₂
Molecular Weight: 269,908 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800918
CAS-Nummer: 951884-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR