N-(4-Methoxybenzyl)-5-brompicolinamid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-N-(4-methoxybenzyl)picolinamide, 5-Bromo-N-(4-methoxybenzyl)pyridine-2-carboxamide

OR11923-250MG
APOSOR11923-250MGEA 34.8 EUR
APOSOR11923-250MG
N-(4-Methoxybenzyl)-5-brompicolinamid
N-(4-Methoxybenzyl)-5-brompicolinamid
Formel: C₁₄H₁₃BrN₂O₂
Molecular Weight: 321.1692 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09801060
CAS-Nummer: 951885-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR