5-Methyl-2-(2-nitrophenyl)oxazol
Lieferant: Apollo Scientific
|
Achtung
|
OR11929-1G
OR11929-100MG
OR11929-500MG
APOSOR11929-1GEA
418
EUR
APOSOR11929-1G
APOSOR11929-100MG
APOSOR11929-500MG
5-Methyl-2-(2-nitrophenyl)oxazol
5-Methyl-2-(2-nitrophenyl)oxazole
Formel:
C₁₀H₈N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09801026 CAS-Nummer: 52829-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...