5-Methyl-2-(2-nitrophenyl)oxazol

Lieferant: Apollo Scientific
Achtung

OR11929-1G OR11929-100MG OR11929-500MG
APOSOR11929-1GEA 418 EUR
APOSOR11929-1G APOSOR11929-100MG APOSOR11929-500MG
5-Methyl-2-(2-nitrophenyl)oxazol
5-Methyl-2-(2-nitrophenyl)oxazole
Formel: C₁₀H₈N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09801026
CAS-Nummer: 52829-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR