4-{[2-(Dimethylamino)ethyl]carbamoyl}benzeneboronic acid hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-((2-(Dimethylamino)ethyl)carbamoyl)phenyl)boronic acid hydrochloride, 4-Borono-N-[2-(dimethylamino)ethyl]benzamide hydrochloride

OR1193-1G
APOSOR1193-1GEA 77 EUR
APOSOR1193-1G
4-{[2-(Dimethylamino)ethyl]carbamoyl}benzeneboronic acid hydrochloride
4-{[2-(Dimethylamino)ethyl]carbamoyl}benzeneboronic acid hydrochloride
Formel: C₁₁H₁₇BN₂O₃·ClH
Molecular Weight: 272,536 g/mol
Schmelzpunkt: 186…190 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06656014
CAS-Nummer: 850568-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR