2-Chlor-5-nitronicotinsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR11941-250MG
APOSOR11941-250MGEA
21.9
EUR
APOSOR11941-250MG
2-Chlor-5-nitronicotinsäure 95%
2-Chlor-5-nitronicotinsäure
Formel:
C₆H₃ClN₂O₄ Schmelzpunkt: 140…143 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658402 CAS-Nummer: 42959-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...