Methylimidazo[1,2-a]pyridin-6-carboxylat Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR11947-250MG
APOSOR11947-250MGEA 29 EUR
APOSOR11947-250MG
Methylimidazo[1,2-a]pyridin-6-carboxylat Hydrochlorid
Methylimidazo[1,2-a]pyridin-6-carboxylat Hydrochlorid
Formel: C₉H₉ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09801003
CAS-Nummer: 957120-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR