2-Amino-5-brom-4-methoxynicotinonitril

Lieferant: Apollo Scientific
Achtung

OR11950-250MG
APOSOR11950-250MGEA 33.5 EUR
APOSOR11950-250MG
2-Amino-5-brom-4-methoxynicotinonitril
2-Amino-5-brom-4-methoxynicotinonitril
Formel: C₇H₆BrN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09801018
CAS-Nummer: 951884-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR