2-(3-Bromphenyl)-1-(pyrrolidin-1-yl)ethanon

Lieferant: Apollo Scientific
Achtung

OR11952-250MG
APOSOR11952-250MGEA 29 EUR
APOSOR11952-250MG
2-(3-Bromphenyl)-1-(pyrrolidin-1-yl)ethanon
2-(3-Bromphenyl)-1-(pyrrolidin-1-yl)ethanon
Formel: C₁₂H₁₄BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09800919
CAS-Nummer: 951884-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR