Ethyl-7-methylimidazo[1,2-a]pyridin-2-carboxylat

Lieferant: Apollo Scientific

OR11957-5G
APOSOR11957-5GEA 154 EUR
APOSOR11957-5G
Ethyl-7-methylimidazo[1,2-a]pyridin-2-carboxylat
Ethyl-7-methylimidazo[1,2-a]pyridin-2-carboxylat
Formel: C₁₁H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02187526
CAS-Nummer: 70705-33-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR