4-(6-Chloropyrazin-2-yl)morpholine 98%

Lieferant: Apollo Scientific
Achtung

OR11963-250MG
APOSOR11963-250MGEA 0 EUR
APOSOR11963-250MG
4-(6-Chloropyrazin-2-yl)morpholine 98%
4-(6-Chloropyrazin-2-yl)morpholine
Formel: C₈H₁₀ClN₃O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 720693-19-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR