N-(tert-Butyl)-6-chloro-4-pyrimidinamine

Lieferant: Apollo Scientific
Achtung

OR11966-250MG
APOSOR11966-250MGEA 29 EUR
APOSOR11966-250MG
N-(tert-Butyl)-6-chloro-4-pyrimidinamine
N-(tert-Butyl)-6-chloro-4-pyrimidinamine
Formel: C₈H₁₂ClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09756959
CAS-Nummer: 945896-38-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR