trans-4-Propylcyclohexancarbonsäure

Lieferant: Apollo Scientific
Achtung

OR11973-5G
APOSOR11973-5GEA 21.9 EUR
APOSOR11973-5G
trans-4-Propylcyclohexancarbonsäure
trans-4-Propylcyclohexanecarboxylic acid
Formel: C₁₀H₁₈O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00059559
CAS-Nummer: 38289-27-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR