5-Chloro-2-methoxy-3-nitropyridine

Lieferant: Apollo Scientific
Achtung

OR11978-5G
APOSOR11978-5GEA 39.3 EUR
APOSOR11978-5G
5-Chloro-2-methoxy-3-nitropyridine
5-Chloro-2-methoxy-3-nitropyridine
Formel: C₆H₅ClN₂O₃
Molecular Weight: 188.569 g/mol
Schmelzpunkt: 90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07375032
CAS-Nummer: 22353-52-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR