4-Bromo-1-isopropylpyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1-isopropyl-1H-pyrazole

OR11985-5G
APOSOR11985-5GEA 59.6 EUR
APOSOR11985-5G
4-Bromo-1-isopropylpyrazole
4-Bromo-1-isopropylpyrazole
Formel: C₆H₉BrN₂
Molecular Weight: 189.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09801021
CAS-Nummer: 313735-62-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR