1-Boc-4-(cyanmethyl)piperidin

Lieferant: Apollo Scientific
Achtung

OR11986-1G
APOSOR11986-1GEA 0 EUR
APOSOR11986-1G
1-Boc-4-(cyanmethyl)piperidin
1-Boc-4-(cyanmethyl)piperidin
Formel: C₁₂H₂₀N₂O₂
Schmelzpunkt: 70…74 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09801019
CAS-Nummer: 256411-39-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR