2-Formyl-6-picolin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Methylpyridine-2-carboxaldehyde, 6-Methyl-2-pyridinecarboxaldehyde
OR12003-25G
APOSOR12003-25GEA
69.6
EUR
APOSOR12003-25G
2-Formyl-6-picolin
2-Formyl-6-picolin
Formel:
C₇H₇NO Molecular Weight: 121.14 g/mol Schmelzpunkt: 31…33 °C Flammpunkt: 68 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006291 CAS-Nummer: 1122-72-1 EINECS: 214-359-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...