4-{2-[4-(Benzyloxy)phenoxy]ethoxy}benzenecarbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR12026-1G
APOSOR12026-1GEA
0
EUR
APOSOR12026-1G
4-{2-[4-(Benzyloxy)phenoxy]ethoxy}benzenecarbaldehyde
4-{2-[4-(Benzyloxy)phenoxy]ethoxy}benzenecarbaldehyde
Formel:
C₂₂H₂₀O₄ Schmelzpunkt: 123…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689765 CAS-Nummer: 937601-87-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...