4-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR12030-1G
OR12030-250MG
APOSOR12030-1GEA
0
EUR
APOSOR12030-1G
APOSOR12030-250MG
4-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde
4-{3-[4-(Benzyloxy)phenoxy]propoxy}benzenecarbaldehyde
Formel:
C₂₃H₂₂O₄ Schmelzpunkt: 92…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689784 CAS-Nummer: 937602-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...