(5-Bromo-2-chloro-3-pyridinyl)methanol

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-chloro-3-(hydroxymethyl)pyridine, (5-Bromo-2-chloropyridin-3-yl)methanol

OR12037-500MG
APOSOR12037-500MGEA 134 EUR
APOSOR12037-500MG
(5-Bromo-2-chloro-3-pyridinyl)methanol
(5-Bromo-2-chloro-3-pyridinyl)methanol
Formel: C₆H₅BrClNO
Molecular Weight: 222,47 g/mol
Schmelzpunkt: 76…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08688594
CAS-Nummer: 742100-75-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR