7-Bromo-2,3-dihydro-1-benzofuran-5-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Bromo-2,3-dihydrobenzo[b]furan-5-carboxaldehyde
OR12038-250MG
APOSOR12038-250MGEA
27.7
EUR
APOSOR12038-250MG
7-Bromo-2,3-dihydro-1-benzofuran-5-carbaldehyde
7-Bromo-2,3-dihydro-1-benzofuran-5-carbaldehyde
Formel:
C₉H₇BrO₂ Molecular Weight: 227.06 g/mol Schmelzpunkt: 109…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689769 CAS-Nummer: 875551-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...