2-Bromoethyl-4-(tert-butyl)phenyl ether

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Bromoethoxy)-4-(tert-butyl)benzene

OR12040-1G
APOSOR12040-1GEA 58 EUR
APOSOR12040-1G
2-Bromoethyl-4-(tert-butyl)phenyl ether
2-Bromoethyl-4-(tert-butyl)phenyl ether
Formel: C₁₂H₁₇BrO
Siedepunkt: 95 °C/0,38mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD02030708
CAS-Nummer: 5952-59-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR