3-Brom-5-nitro-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-5-nitroindazole
OR12046-5G
APOSOR12046-5GEA
21.9
EUR
APOSOR12046-5G
3-Brom-5-nitro-1H-indazol
3-Bromo-5-nitro-1H-indazole
Formel:
C₇H₄BrN₃O₂ Molecular Weight: 242,03 g/mol Schmelzpunkt: 227…229 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07367964 CAS-Nummer: 67400-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...