2-Chloro-1,3-benzothiazole-6-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-6-benzothiazolecarboxylic acid

OR12055-1G
APOSOR12055-1GEA 65.4 EUR
APOSOR12055-1G
2-Chloro-1,3-benzothiazole-6-carboxylic acid
2-Chloro-1,3-benzothiazole-6-carboxylic acid
Formel: C₈H₄ClNO₂S
Molecular Weight: 213,64 g/mol
Schmelzpunkt: 220…221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689736
CAS-Nummer: 3855-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR