{1-[(4-Chlorophenyl)sulphonyl]-4-piperidinyl}methanol

Lieferant: Apollo Scientific
Achtung

OR12068-250MG
APOSOR12068-250MGEA 30.6 EUR
APOSOR12068-250MG
{1-[(4-Chlorophenyl)sulphonyl]-4-piperidinyl}methanol
{1-[(4-Chlorophenyl)sulphonyl]-4-piperidinyl}methanol
Formel: C₁₂H₁₆ClNO₃S
Molecular Weight: 289,78 g/mol
Schmelzpunkt: 102…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689742
CAS-Nummer: 870529-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR