trans-2,5-Dichlorocinnamic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR12077-1G
APOSOR12077-1GEA
46.4
EUR
APOSOR12077-1G
trans-2,5-Dichlorocinnamic acid
trans-2,5-Dichlorocinnamic acid
Formel:
C₉H₆Cl₂O₂ Schmelzpunkt: 192…194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03002786 CAS-Nummer: 20595-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...