6,7-Dimethyl-1,4-dihydro-2,3-quinoxalinedione2-oxime
Lieferant: Apollo Scientific
|
Achtung
|
OR12085-1G
APOSOR12085-1GEA
0
EUR
APOSOR12085-1G
6,7-Dimethyl-1,4-dihydro-2,3-quinoxalinedione2-oxime
6,7-Dimethyl-1,4-dihydro-2,3-quinoxalinedione2-oxime
Formel:
C₁₀H₁₁N₃O₂ Schmelzpunkt: 292…294 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08689715 CAS-Nummer: 937601-74-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...