Ethyl-2-(1-benzyl-4-piperidinylidene)acetate
Lieferant: Apollo Scientific
|
Achtung
|
OR12094-1G
APOSOR12094-1GEA
102
EUR
APOSOR12094-1G
Ethyl-2-(1-benzyl-4-piperidinylidene)acetate
Ethyl-2-(1-benzyl-4-piperidinylidene)acetate
Formel:
C₁₆H₂₁NO₂ Molecular Weight: 259.35 g/mol Siedepunkt: 120…123 °C/3mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437068 CAS-Nummer: 40110-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...