Ethyl-4-[2-(4-hydroxypiperidino)ethoxy]benzenecarboxylate

Lieferant: Apollo Scientific
Achtung

OR12097-1G
APOSOR12097-1GEA 48 EUR
APOSOR12097-1G
Ethyl-4-[2-(4-hydroxypiperidino)ethoxy]benzenecarboxylate
Ethyl-4-[2-(4-hydroxypiperidino)ethoxy]benzenecarboxylate
Formel: C₁₆H₂₃NO₄
Molecular Weight: 293,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08689771
CAS-Nummer: 937601-92-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR