N-Ethyl-2-(methylamino)acetamide hydrochloride

Lieferant: Apollo Scientific
Achtung

OR12098-250MG
APOSOR12098-250MGEA 55.1 EUR
APOSOR12098-250MG
N-Ethyl-2-(methylamino)acetamide hydrochloride
N-Ethyl-2-(methylamino)acetamide hydrochloride
Formel: C₅H₁₃ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD08060121
CAS-Nummer: 909191-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR