5-(Hydroxymethyl)-6-phenyl-4,5-dihydro-3(2H)-pyridazinone

Lieferant: Apollo Scientific
Achtung

OR12101-1G
APOSOR12101-1GEA 92.8 EUR
APOSOR12101-1G
5-(Hydroxymethyl)-6-phenyl-4,5-dihydro-3(2H)-pyridazinone
5-(Hydroxymethyl)-6-phenyl-4,5-dihydro-3(2H)-pyridazinone
Formel: C₁₁H₁₂N₂O₂
Molecular Weight: 204,23 g/mol
Schmelzpunkt: 163…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01692709
CAS-Nummer: 23239-13-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR