4-(Isopentyloxy)benzenecarboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Isopentyloxy)benzoic acid

OR12106-1G
APOSOR12106-1GEA 98.6 EUR
APOSOR12106-1G
4-(Isopentyloxy)benzenecarboxylic acid
4-(Isopentyloxy)benzenecarboxylic acid
Formel: C₁₂H₁₆O₃
Molecular Weight: 208,26 g/mol
Schmelzpunkt: 142…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01814579
CAS-Nummer: 2910-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR