N-Methoxy-N-methyl-2-thiophenecarboxamide

Lieferant: Apollo Scientific
Achtung

OR12108-1G
APOSOR12108-1GEA 21.9 EUR
APOSOR12108-1G
N-Methoxy-N-methyl-2-thiophenecarboxamide
N-Methoxy-N-methyl-2-thiophenecarboxamide
Formel: C₇H₉NO₂S
Molecular Weight: 171,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03643923
CAS-Nummer: 229970-94-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR