Methyl-4-(3-bromopropoxy)benzenecarboxylate

Lieferant: Apollo Scientific
Achtung

OR12129-1G
APOSOR12129-1GEA 92.8 EUR
APOSOR12129-1G
Methyl-4-(3-bromopropoxy)benzenecarboxylate
Methyl-4-(3-bromopropoxy)benzenecarboxylate
Formel: C₁₁H₁₃BrO₃
Molecular Weight: 273,13 g/mol
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01814575
CAS-Nummer: 135998-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR