Methyl-5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate
OR12133-1G
APOSOR12133-1GEA
24.8
EUR
APOSOR12133-1G
Methyl-5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate
Methyl-5,6-dihydro-4H-cyclopenta[b]thiophene-2-carboxylate
Formel:
C₉H₁₀O₂S Molecular Weight: 182,24 g/mol Schmelzpunkt: 60…62 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01927210 CAS-Nummer: 353269-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...