Methyl-4-methyl-2-(methylsulphanyl)-5-pyrimidinecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-methyl-2-(methylthio)pyrimidine-5-carboxylate
OR12143-10G
OR12143-1G
OR12143-5G
APOSOR12143-10GEA
654
EUR
APOSOR12143-10G
APOSOR12143-1G
APOSOR12143-5G
Methyl-4-methyl-2-(methylsulphanyl)-5-pyrimidinecarboxylate
Methyl-4-methyl-2-(methylsulphanyl)-5-pyrimidinecarboxylate
Formel:
C₈H₁₀N₂O₂S Schmelzpunkt: 59…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04125151 CAS-Nummer: 166392-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...