8-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR12151-1G
APOSOR12151-1GEA
40.6
EUR
APOSOR12151-1G
8-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole hydrochloride
8-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole hydrochloride
Formel:
C₁₂H₁₅ClN₂ Molecular Weight: 222,72 g/mol Schmelzpunkt: 270…271 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07781100 CAS-Nummer: 57933-28-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...