1-Benzyl-2-[4-(tert-butyl)phenyl]-6-oxo-3-piperidinecarboxylic acid
Lieferant: Apollo Scientific
OR12169-5G
OR12169-10G
OR12169-1G
APOSOR12169-5GEA
538
EUR
APOSOR12169-5G
APOSOR12169-10G
APOSOR12169-1G
1-Benzyl-2-[4-(tert-butyl)phenyl]-6-oxo-3-piperidinecarboxylic acid
1-Benzyl-2-[4-(tert-butyl)phenyl]-6-oxo-3-piperidinecarboxylic acid
Formel:
C₂₃H₂₇NO₃ Molecular Weight: 365.48 g/mol Schmelzpunkt: 144…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027174 CAS-Nummer: 952182-64-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...