2-Chlor-N-(2,3-dimethylphenyl)acetamid

Lieferant: Apollo Scientific

OR12177-1G
APOSOR12177-1GEA 33.5 EUR
APOSOR12177-1G
2-Chlor-N-(2,3-dimethylphenyl)acetamid
2-Chlor-N-(2,3-dimethylphenyl)acetamid
Formel: C₁₀H₁₂ClNO
Molecular Weight: 197,66 g/mol
Schmelzpunkt: 135…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00018895
CAS-Nummer: 2564-07-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR