5-Bromo-1,2-dimethyl-1H-imidazole 96%
Lieferant: Apollo Scientific
|
Gefahr
|
OR1220-5G
APOSOR1220-5GEA
119
EUR
APOSOR1220-5G
5-Bromo-1,2-dimethyl-1H-imidazole 96%
5-Bromo-1,2-dimethyl-1H-imidazole
Formel:
C₅H₇BrN₂ Molecular Weight: 175.026 g/mol Schmelzpunkt: 91…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659905 CAS-Nummer: 24134-09-6 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...