4-Brom-1,2-dimethylimidazol
Lieferant: Apollo Scientific
|
Achtung
|
OR1222-1G
OR1222-5G
APOSOR1222-1GEA
111
EUR
APOSOR1222-1G
APOSOR1222-5G
4-Brom-1,2-dimethylimidazol
4-Bromo-1,2-dimethylimidazole
Formel:
C₅H₇BrN₂ Schmelzpunkt: 40…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659901 CAS-Nummer: 850429-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...